Tetraethylammonium tetrafluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Tetrylammonium tetrafluoroborate

A902002-10G A902002-25G A902002-5G
76792-078EA 15.92 USD
76792-078 76792-080 76792-082
Tetraethylammonium tetrafluoroborate 97%
Tetraethylammonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₈H₂₀BF₄N
MW: 217.06 g/mol
MDL Number: MFCD00011827
CAS Number: 429-06-1
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR