N-(Azepan-1-ylcarbamoyl)-4-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A899724-100MG A899724-10MG A899724-1G A899724-1MG A899724-250MG A899724-5G A899724-50MG
76792-062EA 53 USD
76792-062 76792-064 76792-066 76792-068 76792-070 76792-072 77873-349
N-(Azepan-1-ylcarbamoyl)-4-methylbenzenesulfonamide 98%
N-(Azepan-1-ylcarbamoyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₃O₃S
MDL Number: MFCD00083504
CAS Number: 1156-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR