4-Amino-5-bromopyrrolo[2,3-d]pyrimidine 95%

Supplier: Ambeed
Warning

A898540-1G A898540-250MG A898540-5G
76792-056EA 58.35 USD
76792-056 76792-058 76792-060
4-Amino-5-bromopyrrolo[2,3-d]pyrimidine 95%
4-Amino-5-bromopyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MDL Number: MFCD09835119
CAS Number: 22276-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR