Tandutinib 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Isopropoxyphenyl)-4-(6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-yl)piperazine-1-carboxamide, CT53518

A895261-100MG A895261-1MG A895261-250MG A895261-50MG
76792-050EA 60.48 USD
76792-050 76792-052 76792-054 77670-310
Tandutinib 98%
Tandutinib

Precaución: for Research Use Only

Formula: C₃₁H₄₂N₆O₄
MW: 562.7 g/mol
MDL Number: MFCD09954147
CAS Number: 387867-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR