4-(Benzyloxy)-3-chlorobenzaldehyde 95%

Supplier: Ambeed
Warning

A893951-1G A893951-250MG A893951-5G
76792-040EA 25 USD
76792-040 76792-042 77603-236
4-(Benzyloxy)-3-chlorobenzaldehyde 95%
4-(Benzyloxy)-3-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
MDL Number: MFCD00831645
CAS Number: 66422-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR