trans-Bis(triphenylphosphine)palladium(II) bromide 98%

Supplier: Ambeed
Warning

Synonyms: trans-Dibromobis(triphenylphosphine)palladium(II)

A874841-1G A874841-250MG A874841-5G
76792-004EA 39 USD
76792-004 76792-006 76792-008
trans-Bis(triphenylphosphine)palladium(II) bromide 98%
trans-Bis(triphenylphosphine)palladium(II) bromide

Precaución: for Research Use Only

Formula: C₃₆H₃₂Br₂P₂Pd
MW: 790.81
MDL Number: MFCD00134175
CAS Number: 22180-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR