Succinic acid dihydrazide 97%

Supplier: Ambeed
Danger

Synonyms: Succinic Dihydrazide

A873783-100G A873783-500G A873783-25G
76792-000EA 15 USD
76792-000 76792-002 76845-556
Succinic acid dihydrazide 97%
Succinic acid dihydrazide

Precaución: for Research Use Only

Formula: C₄H₁₀N₄O₂
MW: 146.1478 g/mol
MDL Number: MFCD00007613
CAS Number: 4146-43-4
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR