TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 98%

Supplier: Ambeed
Warning

Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

A869575-100G A869575-25G A869575-500G A869575-5G
76791-980EA 70.02 USD
76791-980 76791-982 76791-984 76791-986
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 98%
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)

Precaución: for Research Use Only

Formula: C₆H₁₅NO₆S
MW: 229.25 g/mol
MDL Number: MFCD00007532
CAS Number: 7365-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR