Benzofuran-7-carbaldehyde 97%

Supplier: Ambeed
Warning

A868296-100MG A868296-1G A868296-250MG
76791-970EA 19 USD
76791-970 76791-972 77873-328
Benzofuran-7-carbaldehyde 97%
Benzofuran-7-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆O₂
MDL Number: MFCD08272125
CAS Number: 95333-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR