N-Boc-4-(trifluoromethyl)-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-4-trifluoromethyl-L-phenylalanine, Boc-Phe(4-CF3)-OH, N-Boc-4-trifluoromethyl-L-phenylalanine

A868024-100MG A868024-1G A868024-250MG A868024-25G A868024-5G
76791-960EA 9 USD
76791-960 76791-962 76791-964 76791-966 76791-968
N-Boc-4-(trifluoromethyl)-L-phenylalanine 97%
N-Boc-4-(trifluoromethyl)-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MW: 333.31 g/mol
MDL Number: MFCD00792407
CAS Number: 114873-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR