5-amino-2chlorobenzylalcohol 95%

Supplier: Ambeed
Warning

A867720-10G A867720-1G A867720-250MG A867720-5G A867720-25G
76791-952EA 117 USD
76791-952 76791-954 76791-956 76791-958 77873-327
5-amino-2chlorobenzylalcohol 95%
5-amino-2chlorobenzylalcohol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD05664329
CAS Number: 89951-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR