2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid 98%

Supplier: Ambeed
Danger

Synonyms: Suprofen, 2-(4-(Thiophene-2-carbonyl)phenyl)propanoic acid

A867270-10MG A867270-1MG A867270-25MG A867270-50MG A867270-5MG
76791-942EA 23.34 USD
76791-942 76791-944 76791-946 76791-948 76791-950
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid 98%
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃S
MW: 260.30 g/mol
MDL Number: MFCD00079572
CAS Number: 40828-46-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR