4-(4-Methoxyphenyl)cyclohexanone 95%

Supplier: Ambeed
Warning

A863573-100MG A863573-1G A863573-250MG
76791-936EA 32 USD
76791-936 76791-938 76791-940
4-(4-Methoxyphenyl)cyclohexanone 95%
4-(4-Methoxyphenyl)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
MDL Number: MFCD09743528
CAS Number: 5309-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR