4-Bromo-5-fluoro-2-methoxyphenol 95+%

Supplier: Ambeed
Warning

A862008-1G A862008-250MG A862008-5G
76791-930EA 54 USD
76791-930 76791-932 76791-934
4-Bromo-5-fluoro-2-methoxyphenol 95+%
4-Bromo-5-fluoro-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
MDL Number: MFCD08437599
CAS Number: 886510-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR