2,3-Difluoro-4-n-propoxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

A861462-1G A861462-25G A861462-5G
76791-916EA 9 USD
76791-916 76791-918 76791-920
2,3-Difluoro-4-n-propoxybenzeneboronic acid 98%
2,3-Difluoro-4-n-propoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MDL Number: MFCD14584457
CAS Number: 212837-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR