Tyrphostin AG-126 98%

Supplier: Ambeed
Danger

A860936-10MG A860936-1MG A860936-25MG A860936-50MG A860936-5MG
76791-906EA 29 USD
76791-906 76791-908 76791-910 76791-912 76791-914
Tyrphostin AG-126 98%
Tyrphostin AG-126

Precaución: for Research Use Only

Formula: C₁₀H₅N₃O₃
MDL Number: MFCD00236448
CAS Number: 118409-62-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR