Pivalaldehyde (trimethylacetaldehyde) 96% (by GC)

Supplier: Ambeed
Danger

A858364-25G A858364-5G
76791-902EA 85 USD
76791-902 76791-904
Pivalaldehyde (trimethylacetaldehyde) 96% (by GC)
Pivalaldehyde (trimethylacetaldehyde)

Precaución: for Research Use Only

Formula: C₅H₁₀O
MDL Number: MFCD00006962
CAS Number: 630-19-3
UN: 1989
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR