Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Methyl trans-4-(aminomethyl)cyclohexane-1-carboxylate hydrochloride, trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride

A855746-1G A855746-25G A855746-5G
76791-886EA 8.49 USD
76791-886 76791-888 76791-890
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride 98%
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 207.70 g/mol
MDL Number: MFCD00797465
CAS Number: 29275-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR