2-Methoxy-4-methylaniline 96%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-methoxyaniline

A852420-1G A852420-5G
76791-882EA 38.2 USD
76791-882 76791-884
2-Methoxy-4-methylaniline 96%
2-Methoxy-4-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.179 g/mol
MDL Number: MFCD06408246
CAS Number: 39538-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR