Triacetin (glycerol triacetate) 99+%

Supplier: Ambeed
Warning

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

A835553-1000G A835553-100G A835553-25G A835553-500G
76791-824EA 15.92 USD
76791-824 76791-826 76791-828 76791-830
Triacetin (glycerol triacetate) 99+%
Triacetin (glycerol triacetate)

Precaución: for Research Use Only

Formula: C₉H₁₄O₆
MW: 218.21 g/mol
MDL Number: MFCD00008716
CAS Number: 102-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR