Trityl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: Triphenylmethanol, Triphenylcarbinol

A835340-1000G A835340-100G A835340-25G A835340-500G
76791-816EA 69 USD
76791-816 76791-818 76791-820 76791-822
Trityl alcohol 98%
Trityl alcohol

Precaución: for Research Use Only

Formula: C₁₉H₁₆O
MDL Number: MFCD00004445
CAS Number: 76-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR