[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro[(S)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro [(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

A833984-1G A833984-250MG A833984-5G
76791-808EA 38 USD
76791-808 76791-810 77670-196
[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride 98%
[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride

Precaución: for Research Use Only

Formula: C₄₄H₃₄Cl₂P₂Ru
MW: 796.66 g/mol
MDL Number: MFCD01073794
CAS Number: 134524-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR