Fluorescein disodium salt 98% (contains < 6% H₂O)

Supplier: Ambeed
Warning

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

A833092-1000G A833092-25G A833092-500G
76791-802EA 90.18 USD
76791-802 76791-804 76791-806
Fluorescein disodium salt 98% (contains < 6% H₂O)
Fluorescein disodium salt

Precaución: for Research Use Only

Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
MDL Number: MFCD00167039
CAS Number: 518-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR