Pivalic anhydride 98%

Supplier: Ambeed
Danger

Synonyms: Trimethylacetic anhydride, 2,2-Dimethylpropionic anhydride

A830830-25G A830830-5G A830830-100G
76791-798EA 21.99 USD
76791-798 76791-800 77595-324
Pivalic anhydride 98%
Pivalic anhydride

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₃
MDL Number: MFCD00008842
CAS Number: 1538-75-6
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR