3-Fluoro-5-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-5-methylphenyl)methanol

A830457-1G A830457-5G
76791-794EA 39 USD
76791-794 76791-796
3-Fluoro-5-methylbenzyl alcohol 95%
3-Fluoro-5-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD03094314
CAS Number: 518070-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR