5'-Bromo-1',2'-dihydrospiro[cyclohexane-1,3'-indole]-2'-one 97%

Supplier: Ambeed
Warning

Synonyms: 5'-Bromospiro[cyclohexane-1,3'-indolin]-2'-one

A830023-100MG A830023-1G A830023-250MG
76791-788EA 31.83 USD
76791-788 76791-790 76791-792
5'-Bromo-1',2'-dihydrospiro[cyclohexane-1,3'-indole]-2'-one 97%
5'-Bromo-1',2'-dihydrospiro[cyclohexane-1,3'-indole]-2'-one

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO
MW: 280.17 g/mol
MDL Number: MFCD12964079
CAS Number: 304468-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR