6-(Trifluoromethyl)-1H-indazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A829828-100MG A829828-1G A829828-250MG A829828-5G
76791-774EA 31.83 USD
76791-774 76791-776 76791-778 76791-780
6-(Trifluoromethyl)-1H-indazole-3-carboxylic acid 98%
6-(Trifluoromethyl)-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂O₂
MW: 230.14 g/mol
MDL Number: MFCD03840715
CAS Number: 887576-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR