Methyl 3-cyano-4-isopropoxybenzoate 95%

Supplier: Ambeed
Warning

A827925-1G A827925-250MG A827925-5G
76791-764EA 39.26 USD
76791-764 76791-766 76791-768
Methyl 3-cyano-4-isopropoxybenzoate 95%
Methyl 3-cyano-4-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.23 g/mol
MDL Number: MFCD09271837
CAS Number: 213598-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR