Octadecyl methacrylate 96% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
Methacrylic acid Stearyl ester, Octadecyl Methacrylate, Stearyl Methacrylate, Methacrylic acid Octadecyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A827506-100G
A827506-500G
76791-760EA
19
USD
76791-760
76791-762
Octadecyl methacrylate 96% stabilised
Octadecyl methacrylate
Precaución: for Research Use Only
Formula:
C₂₂H₄₂O₂ MW: 338.57 g/mol |
MDL Number:
MFCD00026683 CAS Number: 32360-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...