Tris(trimethylsilyloxy)ethylene 97%

Supplier: Ambeed
Danger

Synonyms: 2,2,7,7-Tetramethyl-4-((trimethylsilyl)oxy)-3,6-dioxa-2,7-disilaoct-4-ene

A825553-1G A825553-250MG A825553-25G A825553-5G
76791-752EA 27 USD
76791-752 76791-754 76791-756 76791-758
Tris(trimethylsilyloxy)ethylene 97%
Tris(trimethylsilyloxy)ethylene

Precaución: for Research Use Only

Formula: C₁₁H₂₈O₃Si₃
MW: 292.60 g/mol
MDL Number: MFCD00011641
CAS Number: 69097-20-7
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR