Zinc sulfate heptahydrate 99%

Supplier: Ambeed
Danger

A824565-25G A824565-500G A824565-100G
76791-750EA 6.37 USD
76791-750 77603-102 77670-172
Zinc sulfate heptahydrate 99%
Zinc sulphate heptahydrate

Precaución: for Research Use Only

Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
MDL Number: MFCD00149894
CAS Number: 7446-20-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR