4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%

Supplier: Ambeed
Warning

Synonyms: Zingerone, Vanillylacetone

A824166-100G A824166-25G A824166-50MG A824166-5G A824166-500G
76791-742EA 110.34 USD
76791-742 76791-744 76791-746 76791-748 77670-170
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00048232
CAS Number: 122-48-5
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR