(2-Benzyloxyphenyl)acetic acid 97%

Supplier: Ambeed
Danger

A823557-1G A823557-250MG A823557-5G
76791-740EA 45 USD
76791-740 77873-290 77873-291
(2-Benzyloxyphenyl)acetic acid 97%
(2-Benzyloxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₅CH₂OC₆H₄CH₂COOH
MDL Number: MFCD00016822
CAS Number: 22047-88-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR