Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate

A823518-250MG A823518-100MG
76791-738EA 172.93 USD
76791-738 77670-168
Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate 95%
Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.1 g/mol
MDL Number: MFCD09878714
CAS Number: 588720-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR