1,3-Propanediamine-N,N,N',N'-tetraacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: Trimethylenediamine-N,N,N',N'-tetraacetic Acid, 2,2',2'',2'''-(Propane-1,3-diylbis(azanetriyl))tetraacetic acid

A822817-25G A822817-5G
76791-734EA 8.49 USD
76791-734 76791-736
1,3-Propanediamine-N,N,N',N'-tetraacetic acid 98%
1,3-Propanediamine-N,N,N',N'-tetraacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₈
MW: 306.27 g/mol
MDL Number: MFCD00210039
CAS Number: 1939-36-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR