Tin (II) trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
A822618-10G
A822618-1G
A822618-25G
A822618-5G
76791-726EA
36.08
USD
76791-726
76791-728
76791-730
76791-732
Tin (II) trifluoromethanesulfonate 98%
Tin(II) trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₂F₆O₆S₂Sn MW: 416.85 g/mol |
MDL Number:
MFCD00191251 CAS Number: 62086-04-8 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...