1-(1-Bromoethyl)-2-fluorobenzene 97%

Supplier: Ambeed
Danger

A821212-100MG A821212-10G A821212-1G A821212-250MG A821212-5G
76791-714EA 39.26 USD
76791-714 76791-716 76791-718 76791-720 76791-722
1-(1-Bromoethyl)-2-fluorobenzene 97%
1-(1-Bromoethyl)-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05 g/mol
MDL Number: MFCD11180278
CAS Number: 405931-46-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR