6-Bromo-4-chlorothieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A820983-100MG A820983-1G A820983-250MG A820983-5G
76791-706EA 26.53 USD
76791-706 76791-708 76791-710 76791-712
6-Bromo-4-chlorothieno[3,2-d]pyrimidine 95%
6-Bromo-4-chlorothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂S
MDL Number: MFCD09746330
CAS Number: 225385-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR