(R)-(+)-1-Boc-2-cyanopyrrolidine 97%

Supplier: Ambeed
Warning

A816889-1G A816889-25G
76791-692EA 41 USD
76791-692 77603-074
(R)-(+)-1-Boc-2-cyanopyrrolidine 97%
(R)-(+)-1-Boc-2-cyanopyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MDL Number: MFCD06411382
CAS Number: 228244-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR