2,6-Di-tert-butyl-p-benzoquinone 98%

Supplier: Ambeed
Warning

A814110-100G A814110-25G A814110-500G A814110-5G
76791-682EA 95 USD
76791-682 76791-684 76791-686 76791-688
2,6-Di-tert-butyl-p-benzoquinone 98%
2,6-Di-tert-butyl-p-benzoquinone

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MDL Number: MFCD00001601
CAS Number: 719-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR