2-Bromo-5-phenylthiazole 96%

Supplier: Ambeed
Warning

A813345-100MG A813345-250MG A813345-1G
76791-680EA 17 USD
76791-680 77603-052 77670-150
2-Bromo-5-phenylthiazole 96%
2-Bromo-5-phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₆BrNS
MDL Number: MFCD11109488
CAS Number: 133311-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR