D-(+)-Sucrose octaacetate 98%

Supplier: Ambeed
Warning

Synonyms: D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

A813260-1000G A813260-100G A813260-25G A813260-500G
76791-672EA 57 USD
76791-672 76791-674 76791-676 76791-678
D-(+)-Sucrose octaacetate 98%
D-(+)-Sucrose octaacetate

Precaución: for Research Use Only

Formula: C₂₈H₃₈O₁₉
MDL Number: MFCD00006623
CAS Number: 126-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR