Methyl 2-cyclohexyl-2-hydroxy-2-phenylacetate 98%

Supplier: Ambeed
Warning

A805356-1G A805356-25G A805356-5G
76791-646EA 57 USD
76791-646 76791-648 77873-272
Methyl 2-cyclohexyl-2-hydroxy-2-phenylacetate 98%
Methyl 2-cyclohexyl-2-hydroxy-2-phenylacetate

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
MW: 248.31 g/mol
MDL Number: MFCD00269807
CAS Number: 10399-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR