Stearyl stearate 98%

Supplier: Ambeed
Warning

Synonyms: Stearic acid stearyl ester, Octadecyl stearate, Octadecyl octadecanoate

A804449-100G A804449-25G A804449-500G
76791-636EA 24 USD
76791-636 76791-638 76791-640
Stearyl stearate 98%
Stearyl stearate

Precaución: for Research Use Only

Formula: C₃₆H₇₂O₂
MW: 536.97 g/mol
Melting Pt: 64
MDL Number: MFCD00056225
CAS Number: 2778-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR