2-Bromo-5-iodothiazole 95%

Supplier: Ambeed
Danger

A799701-1G A799701-5G
76791-628EA 30 USD
76791-628 77873-265
2-Bromo-5-iodothiazole 95%
2-Bromo-5-iodothiazole

Precaución: for Research Use Only

Formula: C₃HBrINS
MDL Number: MFCD09999190
CAS Number: 108306-63-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR