4'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

A799122-1G A799122-250MG A799122-5G
76791-622EA 44.56 USD
76791-622 76791-624 76791-626
4'-(Trifluoromethyl)biphenyl-4-carboxylic acid 98%
4'-(Trifluoromethyl)biphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
MDL Number: MFCD03424606
CAS Number: 195457-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR