5-(Bromomethyl)-2-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
A793147-1G
A793147-250MG
76791-594EA
119.89
USD
76791-594
76791-596
5-(Bromomethyl)-2-(trifluoromethyl)pyridine 98%
5-(Bromomethyl)-2-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₇H₅BrF₃N MW: 240.021 g/mol |
MDL Number:
MFCD10697684 CAS Number: 108274-33-5 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...