2,2,4,4,6,6-Hexamethyl-1,3,5-trithiane 98%

Supplier: Ambeed
Danger

Synonyms: Trithioacetone

A790602-1G A790602-25G A790602-5G
76791-588EA 5.31 USD
76791-588 76791-590 76791-592
2,2,4,4,6,6-Hexamethyl-1,3,5-trithiane 98%
2,2,4,4,6,6-Hexamethyl-1,3,5-trithiane

Precaución: for Research Use Only

Formula: C₉H₁₈S₃
MW: 222.42 g/mol
MDL Number: MFCD00066341
CAS Number: 828-26-2
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR