Ethyl 4,4,4-trifluoro-3-(methylamino)but-2-enoate 95%

Supplier: Ambeed
Danger

A780538-1G A780538-25G A780538-5G
76791-570EA 15.92 USD
76791-570 76791-572 76791-574
Ethyl 4,4,4-trifluoro-3-(methylamino)but-2-enoate 95%
Ethyl 4,4,4-trifluoro-3-(methylamino)but-2-enoate

Precaución: for Research Use Only

Formula: C₇H₁₀F₃NO₂
MW: 197.15 g/mol
MDL Number: MFCD01075682
CAS Number: 121303-76-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR