1-Boc-5-methoxyindole 98%

Supplier: Ambeed
Danger

A776266-1G A776266-250MG A776266-25G A776266-5G
76791-544EA 16 USD
76791-544 76791-546 76791-548 76791-550
1-Boc-5-methoxyindole 98%
1-Boc-5-methoxyindole

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD08272248
CAS Number: 99275-47-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR